Learn More
CD3E, Human, Matched Antibody Pair (AP41), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00000916-AP41.1Set
Product Code. 16053738
  • 809.00 € / 1 set
Explore more special offers
Includes: Capture antibody: mouse monoclonal anti-CD3E (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CD3E, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
CD3E
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CD3E.
FLJ18683/T3E/TCRE
Mouse
RUO
916
Human
Liquid
ELISA
Biotin
CD3E
CD3E
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CD3E (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CD3E, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.