missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD45; Clone: HI30; BD
363.00 € - 760.00 €
Specifications
Antigen | CD45 |
---|---|
Clone | HI30 |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 395 |
Host Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15833099
|
BD Biosciences
563791 |
25 Tests |
363.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15843099
|
BD Biosciences
563792 |
100 Tests |
760.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Host Species: Mouse
Clone: HI30
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human Peripheral Blood Leucocytes
Flow cytometry
Specifications
CD45 | |
Monoclonal | |
Mouse | |
Human | |
Primary |
HI30 | |
Brilliant Ultraviolet 395 | |
RUO | |
Human Peripheral Blood Leucocytes | |
Affinity Purified |