Learn More
anti-CTGF, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00001490-AP41.1Set
Product Code. 16004338
  • 809.00 € / 1 set
Explore more special offers
Includes: Capture antibody: mouse monoclonal anti-CTGF, IgG2b Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-CTGF, IgG2b Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
CTGF
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CTGF.
CCN2/HCS24/IGFBP8/MGC102839/NOV2
Mouse
RUO
1490
Human
Liquid
ELISA
Biotin
CTGF
CTGF
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CTGF, IgG2b Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-CTGF, IgG2b Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.