missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-DVL1, CTNNB1 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0573.1Set
Includes: DVL1 rabbit purified polyclonal antibody (20μg); CTNNB1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the DVL1 protein, and the other against the CTNNB1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
DVL1 | |
Monoclonal/Polyclonal | |
DVL1 | |
DVL1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. DVL1 rabbit purified polyclonal antibody (20 ug) 2. CTNNB1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
DVL/MGC54245 | |
Mouse, Rabbit | |
RUO | |
1855/1499 | |
Human | |
DVL1 & CTNNB1 Protein Protein Interaction Antibody Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu