missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-F7, F3 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0178.1Set
Includes: F7 rabbit purified polyclonal antibody (20μg); F3 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the F7 protein, and the other against the F3 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
F7 | |
Monoclonal/Polyclonal | |
F7 | |
Mouse, Rabbit | |
RUO | |
2155/2152 | |
Human | |
F7 & F3 Protein Protein Interaction Antibody Pair |
In situ PLA | |
Unconjugated | |
F7 | |
1 Set | |
Primary | |
Antibody pair set content: 1. F7 rabbit purified polyclonal antibody (20 ug) 2. F3 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction