Learn More
Flotillin-1 Mouse, Unlabeled, Clone: 18, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610820
Description
Caveolae are specialized membrane invaginations of 50-100 nm present in all cells, but abundant in endothelium, muscle cells, and adipocytes. These plasma membrane microdomains function in transcytosis of macromolecules, and are the sites of potocytosis, where small molecules are concentrated and transferred inside the cells by glycosylphosphatidylinositol (GPI)-linked receptors. Caveolin, a 22kDa protein and a well known marker for these plasma membrane microdomains, plays a structural role in these specializations. Flotillin-1 was isolated from the Triton X-100 insoluble buoyant fraction, characteristic of caveolae. Although the mRNA expression of both Flotillin-1 and Caveolin
is very similar, Caveolin is undetectable in brain, while Flotillin-1 is very abundant. Flotillin-1 is a close homolog of the Epidermal Surface Antigen (ESA/Flotillin-2), which also colocalizes in the caveolae. Thus, Flotillin-1 and its relative ESA/Flotillin-2 are now incorporated into the expanding list of proteins co-localized at the caveolae which includes PKCα, Ras, Rap Src-like kinases, Gαβγ,and GPI-linked receptors.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
Flotillin-1 | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Mouse Flotillin aa. 312-428 | |
50 μg | |
Cell Biology | |
Chicken, Human, Mouse, Rat | |
IgG1 |
Western Blot | |
18 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.