Learn More
Flotillin-1 Mouse, Unlabeled, Clone: 18, BD
Mouse Monoclonal Antibody
436.00 € - 695.00 €
Specifications
Antigen | Flotillin-1 |
---|---|
Clone | 18 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15835659
|
BD Biosciences
610820 |
50 μg |
436.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15845659
|
BD Biosciences
610821 |
150 μg |
695.00 €
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Caveolae are specialized membrane invaginations of 50-100 nm present in all cells, but abundant in endothelium, muscle cells, and adipocytes. These plasma membrane microdomains function in transcytosis of macromolecules, and are the sites of potocytosis, where small molecules are concentrated and transferred inside the cells by glycosylphosphatidylinositol (GPI)-linked receptors. Caveolin, a 22kDa protein and a well known marker for these plasma membrane microdomains, plays a structural role in these specializations. Flotillin-1 was isolated from the Triton X-100 insoluble buoyant fraction, characteristic of caveolae. Although the mRNA expression of both Flotillin-1 and Caveolin
is very similar, Caveolin is undetectable in brain, while Flotillin-1 is very abundant. Flotillin-1 is a close homolog of the Epidermal Surface Antigen (ESA/Flotillin-2), which also colocalizes in the caveolae. Thus, Flotillin-1 and its relative ESA/Flotillin-2 are now incorporated into the expanding list of proteins co-localized at the caveolae which includes PKCα, Ras, Rap Src-like kinases, Gαβγ,and GPI-linked receptors.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
Flotillin-1 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
18 | |
Western Blot | |
Unconjugated | |
RUO | |
Chicken, Human, Mouse, Rat | |
Mouse Flotillin aa. 312-428 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.