missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-GAB1, CRKL Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0106.1Set
Includes: GAB1 rabbit purified polyclonal antibody (20μg); CRKL mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the GAB1 protein, and the other against the CRKL protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
GAB1, CRKL | |
Unconjugated | |
1 Set | |
Primary | |
Purified |
Monoclonal/Polyclonal | |
Mouse, Rabbit | |
RUO | |
Human |