missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Human IgG, peroxidase-linked species-specific whole antibody (from sheep) Secondary Antibody, Cytiva
Brand: Life Science products Cytiva NA933-1ML
629.85 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
Host Species: Sheep
Isotype: IgG
Species Reactivity: Hu
Protein Blotting, ELISA, Immunocytochemistry
Specifications
IgG | |
HRP | |
Affinity Purified | |
Secondary |
Monoclonal | |
Sheep | |
1 mL | |
IgG |
Safety and Handling
Signal Word
Hazard Category
- Warning
Hazard Category
- Respiratory sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction