missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1, AmCyan, Clone: X40, BD
Brand: BD Biosciences 339185
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: X40
Isotype: IgG1, κ
Cross Reactivity: Human
Control, Flow Cytometry
Specifications
Flow Cytometry | |
AmCyan | |
Mouse | |
100 Tests | |
Isotype Control (Primary) | |
IgG1 κ |
100μg/mL | |
Phosphate buffered saline with BSA and 0.1% sodium azide. | |
Keyhole limpet hemocyanin (KLH) | |
RUO | |
Store vials at 2°C-8°C. Conjugated forms should not be frozen. Protect from exposure to light. Each reagent is stable until the expiration date shown on the bottle label when stored as directed. |