missing translation for 'onlineSavingsMsg'
Learn More

anti-IgG2, κ, Clone: B81-3, BD

297.00 € - 860.00 €

Specifications

Clone B81-3
Applications Blocking Assay, Functional Assay, Cytotoxicity Assay
Conjugate Unconjugated
Host Species Hamster
Isotype IgG2 κ
View More Specs

Products 2
Product Code Brand Concentration Price Quantity & Availability  
Product Code Brand Concentration Price Quantity & Availability  
15819698
View Documents
BD Biosciences
559277
0.5mg/mL
297.00 €
Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
15829718
View Documents
BD Biosciences
559339
1mg/mL
860.00 €
Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
Description

Description

The B81-3 antibody is specific for keyhole limpet hemocyanin. This antibody is routinely tested by LAL assay for endotoxin levels. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Blocking Assay, Co-Stimulation, Cytotoxicity Assay, Isotype Control

Specifications

Specifications

B81-3
Unconjugated
IgG2 κ
Undiluted at 4°C
Blocking Assay, Functional Assay, Cytotoxicity Assay
Hamster
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications