missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG2a, PE, Clone: X39, BD
Brand: BD Biosciences 349053
This item is not returnable.
View return policy
Description
Flow Cytometry
Specifications
Flow Cytometry | |
PE | |
Affinity Purified | |
Isotype Control (Primary) | |
Undiluted at 2° to 8°C. Protect from exposure to light. Do not freeze. |
X39 | |
Mouse | |
100 Tests | |
Human | |
IgG2a κ |