missing translation for 'onlineSavingsMsg'
Learn More

IL-12 (p40) Mouse anti-Human, Unlabeled, Clone: C8.3, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 551227

408.68 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15856578

  • 409.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The C8.3 antibody reacts with human IL-12 p40 subunit but not IL-12 p70 heterodimer or IL-12 p35 monomer. The immunogen used to generate the C8.3 hybridoma was the purified p40 subunit of human IL-12. The C8.3 antibody has been reported not to bind p40 chain when it is complexed to p35 (i.e., the p70 heterodimer). The combination of anti-p40 antibodies, C8.3 and C8.6 , has been reported to yield an assay for detecting only free p40 chain, and specifically not the p70 heterodimer. It has been reported that activated PBMC produce an excess of the
free p40 protein over the p70 heterodimer.

Host Species: Mouse
Clone: C8.3
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified human IL-12 p40 subunit

ELISA

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-12 (p40)
Monoclonal
1.0mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Purified human IL-12 p40 subunit
1 mg
Immunology
Human
IgG1 κ
ELISA
C8.3
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.