Learn More
IL-12 (p40/p70) Mouse anti-Human, Biotin, Clone: C8.6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554660
785.97 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The C8.6 antibody reacts with human IL-12 p40 monomers, homodimers, and p70 heterodimers, but not p35 subunits. The immunogen used to generate the C8.6 hybridoma was the purified p40 subunit of human IL-12. The C8.6 antibody is a neutralizing antibody and has been reported to strongly inhibit three different biological activities of IL-12 (i.e., IFN-γ induction), mitogenic effects on PHA blasts, and enhancement of NK cell-mediated cytotoxicity. C8.6 has been shown to immunoprecipitate IL-23. This antibody is routinely tested by ELISA as a detection antibody. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: C8.6
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified p40 Subunit of Human IL-12
ELISA
Specifications
IL-12 (p40/p70) | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Purified p40 Subunit of Human IL-12 | |
0.5 mg | |
Immunology | |
Human | |
IgG1 κ |
ELISA | |
C8.6 | |
Biotin | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.