Learn More
IL-12 (p40/p70) Mouse anti-Human, Brilliant Violet 421, Clone: C8.6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565023
410.88 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The C8.6 antibody reacts with human IL-12 p40 monomers, homodimers, and p70 heterodimers, but not p35 subunits. The immunogen used to generate the C8.6 hybridoma was the purified p40 subunit of human IL-12. The C8.6 antibody is a neutralizing antibody and has been reported to strongly inhibit three different biological activities of IL-12 (i.e., IFN- γinduction), mitogenic effects on PHA blasts, and enhancement of NK cell-mediated cytotoxicity. C8.6 has been shown to immunoprecipitate IL-23.
The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifications
IL-12 (p40/p70) | |
Monoclonal | |
Brilliant Violet 421 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
Purified p40 Subunit of Human IL-12 | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
C8.6 | |
IL12B; IL-12B; IL12p40; IL-12-40; CLMF; CLMF2; NKSF; NKSF2 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.