missing translation for 'onlineSavingsMsg'
Learn More

IL-12 (p40/p70) Mouse anti-Human, NA/LE, Unlabeled, Clone: C8.6, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 554659

336.41 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15837828

  • 337.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The C8.6 antibody reacts with human IL-12 p40 monomers, homodimers, and p70 heterodimers, but not p35 subunits. The immunogen used to generate the C8.6 hybridoma was the purified p40 subunit of human IL-12. The C8.6 antibody is a neutralizing antibody and has been reported to strongly inhibit three different biological activities of IL-12 (i.e., IFN-γ induction), mitogenic effects on PHA blasts, and enhancement of NK cell-mediated cytotoxicity. C8.6 has been shown to immunoprecipitate IL-23. This antibody is routinely tested by neutralization. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: C8.6
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified p40 Subunit of Human IL-12

Functional Assay, Neutralization

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-12 (p40/p70)
Monoclonal
1.0mg/mL
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Purified p40 Subunit of Human IL-12
0.5 mg
Immunology
Human
IgG1 κ
ELISA
C8.6
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.