missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-JUP, ERBB2 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0160.1Set
Includes: JUP rabbit purified polyclonal antibody (20μg); ERBB2 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the JUP protein, and the other against the ERBB2 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
JUP | |
Monoclonal/Polyclonal | |
JUP | |
JUP | |
1 Set | |
Primary | |
Antibody pair set content: 1. JUP rabbit purified polyclonal antibody (20 ug) 2. ERBB2 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
ARVD12/CTNNG/DP3/DPIII/PDGB/PKGB | |
Mouse, Rabbit | |
RUO | |
3728/2064 | |
Human | |
JUP & ERBB2 Protein Protein Interaction Antibody Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction