Learn More
Ambion™ Anti-miR™ miRNA Inhibitor
Anti-miR™ miRNA Inhibitor
Brand: Ambion™ AM17000
221.77 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.
Specifications
The Anti-miR™ miRNA is provided dried down in a single tube, along with Nuclease-free Water for resuspension, and should be stored at –20°C. It is guaranteed for 1 year when stored dried at –20°C. Keep in a dry, cool and well-ventilated place | |
Anti-miR™ miRNA Inhibitor | |
Ambion™, Anti-miR™ | |
5 nmol | |
Powder |
Anti-miR™ miRNA Inhibitor, 5nmol, Solid, Powder, Keep in a Dry, Cool and Well-Ventilated Place | |
1 Each | |
Anti-miR™ miRNA Inhibitor | |
5nmol |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Ambion™ Anti-miR™ miRNA Inhibitor > 5 nmol
For Research Use Only. Not for use in diagnostic procedures.
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu