Learn More
Ambion™ Anti-miR™ miRNA Inhibitor
Anti-miR™ miRNA Inhibitor
Brand: Ambion™ AM17001
755.76 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically bind to and inhibit endogenous miRNA molecules and enable miRNA functional analysis by down-regulation of miRNA activity.Specifications
Room temperature | |
Custom Anti-miR™ miRNA Inhibitor, 20nmol, Powder, Small, Chemically Modified Single-Stranded RNA Molecules, Designed to Specifically Bind to and Inhibit Endogenous miRNA, Molecules and Enable miRNA Functional Analysis by Down-Regulation of miRNA Activity | |
Custom Anti-miR™ miRNA Inhibitor | |
20 nmol |