Learn More
Novus Biologicals™ Anti-Mouse Ig (H+L) Comp-Bead 3 Population (5.5 um) Kit
The Anti-Mouse Ig (H+L) Compensation Beads are designed to capture fluorophore-conjugated antibodies, serving as accurate and consistent compensation controls in multicolor flow cytometry.
Brand:  Novus Biologicals™ NBP3-00497
Product Code. 18300057
  • 426.55 € / 1 set
Explore more special offers
Specifications
Store at 4°C in the dark. Do not freeze.
3 x 5 mL
PBS, 0.2% BSA
Flow Cytometry