missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Mouse IgG peroxidase-linked whole antibody (from sheep) Secondary Antibody, Cytiva
Highly species specific HRP-conjugated antibodies optimized for use with Amersham ECL Western Blotting Detection Reagents.
Brand: Life Science products Cytiva NXA931-1ML
This item is not returnable.
View return policy
Description
Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms
Protein Blotting, ELISA, Immunocytochemistry

Specifications
IgG, Whole Molecule | |
HRP | |
Affinity Purified | |
Secondary |
Monoclonal | |
Sheep | |
1 mL | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction