missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-NFKB1, STAT3 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0372.1Set
Includes: NFKB1 rabbit purified polyclonal antibody (20μg); STAT3 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the NFKB1 protein, and the other against the STAT3 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
NFKB1 | |
Monoclonal/Polyclonal | |
NFKB1 | |
NFKB1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. NFKB1 rabbit purified polyclonal antibody (20 ug) 2. STAT3 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
DKFZp686C01211/EBP-1/KBF1/MGC54151/NF-kappa-B/NFKB-p105/NFKB-p50/p105/p50 | |
Mouse, Rabbit | |
RUO | |
4790/6774 | |
Human | |
NFKB1 & STAT3 Protein Protein Interaction Antibody Pair |