missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-PRKCZ, F11R Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0171.1Set
Includes: PRKCZ rabbit purified polyclonal antibody (20μg); F11R mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the PRKCZ protein, and the other against the F11R protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
PRKCZ | |
Monoclonal/Polyclonal | |
PRKCZ | |
PRKCZ | |
1 Set | |
Primary | |
Antibody pair set content: 1. PRKCZ rabbit purified polyclonal antibody (20 ug) 2. F11R mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
PKC-ZETA/PKC2 | |
Mouse, Rabbit | |
RUO | |
5590/50848 | |
Human | |
PRKCZ & F11R Protein Protein Interaction Antibody Pair |