missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-PSMD9, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand: Abnova H00005715-AP22.1Set
Includes: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-PSMD9 (100μg) , Detection antibody: mouse purified polyclonal anti-PSMD9 (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
PSMD9 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PSMD9. | |
MGC8644/Rpn4/p27 | |
Mouse, Rabbit | |
RUO | |
5715 | |
Human | |
Liquid |
ELISA | |
Unconjugated | |
PSMD9 | |
PSMD9 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PSMD9 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-PSMD9 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only