missing translation for 'onlineSavingsMsg'
Learn More

anti-Rabbit IgG, biotinylated speciesspecific whole antibody (from donkey), Cytiva

Brand:  Life Science products Cytiva RPN1004-2ML

485.45 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


Product Code. 10460705

  • 526.00 € / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Host Species: Donkey
Isotype: IgG
Species Reactivity: Rb

Protein Blotting, ELISA, Immunocytochemistry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG, Whole Molecule
Biotin
Affinity Purified
Primary
Monoclonal
Donkey
2 mL
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers