missing translation for 'onlineSavingsMsg'
Learn More

anti-Rat IgG, Horseradish Peroxidase-Linked Species-Specific Whole Antibody (from goat) Secondary Antibody, Cytiva

Use for protein blotting, ELISA and immunocytochemistry applications. Anti-Rat IgG, Horseradish Peroxidase-Linked Species-Specific Whole Antibody (from goat) Secondary Antibody, Cytiva, is a secondary antibody from goat.

Brand:  Life Science products Cytiva NA935

539.60 EUR valid until 2025-12-31
Use promo code "24280" to get your promotional price.



Product Code. 10360075

  • 568.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Rt

Protein Blotting, ELISA, Immunocytochemistry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG
HRP
Affinity Purified
Secondary
Monoclonal
Goat
1 mL
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • ||Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : reaction mass of: 5-chloro-2-methyl-4-isothiazolin-3-one ¢ECno. 247-500-7! and2-methyl-2H-isothiaz
  • UNKNOWN ACUTE-4 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.