missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2b, Pacific Orange™, Isotype Control, Invitrogen™

Rat Isotype Control

Brand:  Invitrogen R2B30

Product Code. 10159812

  • 241.00 € / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rat IgG2b
100 ug/ml
PBS with 4mg/mL BSA and 0.1% sodium azide
Rat
Purified
RUO
None
Liquid
Flow Cytometry
Pacific Orange
IgG2, IgG2b
Rat IgG2b
500 μL
Isotype Control (Primary)
Store at 4°C, in the dark
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers