missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS3, Human, Matched Antibody Pair (AP11), Abnova™
Brand: Abnova H00005998-AP11.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-RGS3 (100μg); Detection antibody: mouse monoclonal anti-RGS3, IgG1 Kappa (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RGS3.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifications
RGS3 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human RGS3. | |
C2PA/FLJ20370/FLJ31516/FLJ90496/PDZ-RGS3/RGP3 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-RGS3 (100 μg) 2. Detection antibody: mouse monoclonal anti-RGS3, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
RGS3 | |
RGS3 | |
Affinity Purified | |
RUO | |
5998 | |
Human |