Learn More
Brady™ Anti-skid Laminated Polyester Tape with Stripes
Anti-Skid (anti-slip) Laminated Polyester Tape with Stripes helps to prevent slipping and falling on wet or slippery floors.
Brand:  Brady™ 078146
 View more versions of this product
Product Code. 11751645
  • 79.35 € / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The rugged polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit.
  • Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors.
  • Anti-Skid tape is available in a variety of colors and sizes to suit all of your floor marking needs.
Specifications
Specifications
ANSI A1264.2-2006, NSFI Certified for High Traction, OSHA 1910.144, OSHA 1910.22
18.29 m x 25.40 mm
60 ft.
1 Roll
-40°C to +70°C
1 in.
Permanent Acrylic
Polyester
Striped Anti-Skid Tape - Black / Yellow
Traction on steps, slick floors, and egress pathway marking
18.29 m
-40°F to +158°F
0.162 mm
25.4 mm
Black and Yellow Striped
Anti-skid Tape