missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Smooth Muscle Actin Monoclonal Antibody (1A4), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA106110

Product Code. 11528782

  • 520.00 € / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MA1-06110 detects a-smooth muscle actin (a-SM1) in human, rat, goat, porcine, non-human primate, quail, ovine, xenopus, monkey and chicken samples. Since the epitope recognized by MA1-06110 is highly conserved, it can also cross-react with protochordates, lower craniates and mammals other than those specifically listed. MA1-06110 has sucessfully been used in immunocytochemistry, immunomicroscopy, immunohistochemistry, and Western blotting procedures. The MA1-06110 immunogen is a peptide corresponding to residues E(1) E E D S T A L V C(10) of alpha-smooth muscle actin with an acetylated n-terminus coupled to KLH. MA1-06110 specifically recognizes the epitope Ac-EEED on alpha-smooth muscle actin.

Smooth Muscle Actin belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actin being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. In particular, smooth muscle actin is an alpha actin that is found in skeletal muscle. Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. Smooth muscle actin is found on smooth muscle vessel walls, gut wall, myometrium, myoepithelial cells in breast and salivary glands. Defects in the smooth muscle actin gene cause aortic aneurysm familial thoracic type 6. Actin isoforms differ slightly in their N-terminus and the sequences of each are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown that smooth muscle actin appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases. Multiple alternatively spliced variants of smooth muscle actin have been identified.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Alpha-Smooth Muscle Actin
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P08023, P62736, P62737, P62738
ACTA2, acta2.L
Peptide corresponding to residues E(1) E E D S T A L V C(10) of alpha-smooth muscle actin with an acetylated n-terminus coupled to KLH.
50 μg
Primary
Avian, Chicken, Goat, Human, Mouse, Non-human Primate, Ovine, Porcine, Rat, X. laevis
Antibody
IgG2a
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot
1A4
Unconjugated
ACTA2
0610041G09Rik; AAT6; ACT-4; acta2; acta2.L; acta2.S; actin; Actin Alpha 2 Smooth Muscle Aorta; actin alpha 2, smooth muscle; Actin Vascular Smooth Muscle; actin, alpha 2, smooth muscle, aorta; actin, alpha 2, smooth muscle, aorta L homeolog; actin, alpha 2, smooth muscle, aorta S homeolog; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; ACTSA; ACTVS; alpha actin 2; Alpha Cardiac Actin; alpha smooth muscle actin; alpha-actin; Alpha-actin-2; alpha-cardiac actin; alphaSMA; alpha-SM-actin; alpha-smooth muscle actin; Aortic Smooth Muscle; ASMA; a-sma; cell growth-inhibiting gene 46 protein; GIG46; Growth Inhibiting Gene 46; hypothetical protein LOC515610; LOW QUALITY PROTEIN: actin, aortic smooth muscle; mymy5; SM actin; sma; SMactin; sm-actin; SMalphaA; smooth muscle; smooth muscle alpha-actin; vascular smooth muscle alpha-actin; wu:fb63d03; XELAEV_18034370mg
Mouse
purified
RUO
101121051, 102191486, 11475, 423787, 431847, 59, 733615, 81633
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers