Learn More
Sartorius Anti-Static Devices for Balances
Control charges on samples
1950.00 €
Specifications
For Use With (Equipment) | CPA Balance, Extended Balance, SE and ME Ultramicro Balance |
---|
Description
- Anti-static pen—direct the pen at item to be neutralized
- Ionizing blower—point blower toward sample prior to weighing
Specifications
CPA Balance, Extended Balance, SE and ME Ultramicro Balance |