missing translation for 'onlineSavingsMsg'
Learn More

Sulfaguanidine Polyclonal Antibody, Invitrogen™

Sheep Polyclonal Antibody

Brand:  Invitrogen PA175427

Product Code. 11599030

  • 684.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

PA1-75427 specifically detects Sulfaguanidine. PA1-75427 has been successfully used in ELISA applications. The PA1-75427 immunogen is Sulguanidine-BSA.

Sulfaguanidine is a guanidine derivative of sulfanilamide used in veterinary medicine. Sulfaguanidine is poorly absorbed from the gut but is well suited for the treatment of bacillary dysentery and other enteric infections. [National Center for Biotechnology Information. PubChem Compound Database; CID=5324].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sulfaguanidine
Polyclonal
PBS with 0.09% sodium azide
Sheep
purified
RUO
Chemical
Antibody
IgG
ELISA
Unconjugated
C7H10N4O2S; Guanicil; Sulfaguanidin; Sulfaguine; Sulphaguanidine
Sulguanidine-BSA
1 mg
Primary
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers