missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) chloride, ACS, 99.0% min, Thermo Scientific Chemicals

Product Code. 11373348
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols and carotenoids. It is likewise employed as a dye mordant and a fabric flame retardant. It is used as inorganic and organic chlorination catalyst. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 223.5°C
Melting Point 73.4°C
Color White
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99% min.
Solubility Information Soluble in water,HCl solution,alcohol,benzene,carbon disulfide,dioxane,chloroform,ether,acetone,carbon tetrachloride. Insoluble in organic bases.
Formula Weight 228.11
Grade ACS
Odor Pungent
Sensitivity Moisture sensitive
Appearance White crystalline
Density 3.14 g/cm3
Chemical Name or Material Antimony(III) chloride, For analysis ACS
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'