missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) iodide, ultra dry, 99.998% (metals basis)

Product Code. p-7050124
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11495040

Brand: Thermo Scientific Alfa Aesar 035676.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-44-5 | I3Sb | 502.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-44-5
Molecular Formula I3Sb
Molecular Weight (g/mol) 502.47
MDL Number MFCD00016322
InChI Key KWQLUUQBTAXYCB-UHFFFAOYSA-K
Synonym antimony iii iodide, stibine, triiodo, antimony triiodide, antimon iii iodid, antimony iodide sbi3, antimony iodide, antimony iii iodide, ultra dry, antimony iii triiodide, hsdb 437, triiodostibine #
PubChem CID 24630
SMILES [Sb+3].[I-].[I-].[I-]

Specifications

Boiling Point 420°C
Melting Point 168°C
Physical Form Beads
Quantity 1 g
UN Number UN3260
Merck Index 14,710
Solubility Information Decomposed by water to SbOI.
IUPAC Name triiodostibane
Formula Weight 502.46
Percent Purity 99.998%
Odor Acrid
Packaging Ampule
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Antimony(III) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.