missing translation for 'onlineSavingsMsg'
Learn More
Antimony(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11340038

Antimony(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11340038
10 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11340038

Brand: Thermo Scientific Alfa Aesar 042831.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antimony(III) iodide is used as a dopant in the preparation of thermoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-44-5
Molecular Formula I3Sb
Molecular Weight (g/mol) 502.47
MDL Number MFCD00016322
InChI Key KWQLUUQBTAXYCB-UHFFFAOYSA-K
Synonym antimony iii iodide, stibine, triiodo, antimony triiodide, antimon iii iodid, antimony iodide sbi3, antimony iodide, antimony iii iodide, ultra dry, antimony iii triiodide, hsdb 437, triiodostibine #
PubChem CID 24630
IUPAC Name triiodostibane
SMILES [Sb+3].[I-].[I-].[I-]

Specifications

Boiling Point 420°C
Melting Point 168°C
Physical Form Powder
Quantity 10 g
UN Number UN3260
Merck Index 14,710
Solubility Information Soluble in benzene,alcohol,acetone,CS2,HCl,KI,SnCl4,C2H7N. Insoluble in CHCl3,and CCl4.Soluble in benzene,alcohol,acetone,carbondisulfide,hydrochloric acid,potassium iodide,tinterachloride,dimethylamine. Slightly soluble in ethanol and acetone. Insoluble in chloroform and carbonterachloride.
Formula Weight 502.46
Percent Purity 99.999%
Odor Acrid
Packaging Ampule
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Antimony (III) iodide, Ultra dry
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.