missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) methoxide, 95%, Thermo Scientific™

Product Code. 15451327
Click to view available options
Quantity:
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 29671-18-9
Melting Point 124°C
Flash Point >110°C (230°F)
Molecular Formula C3H9O3Sb
MDL Number MFCD00014882
Quantity 2 g
UN Number UN3181
Sensitivity Air and moisture sensitive
Synonym antimony iii methoxide, acmc-20alrv, antimony 3+ trimethoxide, antimony 3+ tris methoxide, antimony 3+ ion tris methoxide
InChI Key KEUYHGXCOWNTEJ-UHFFFAOYSA-N
SMILES C[O-].C[O-].C[O-].[Sb+3]
IUPAC Name antimony(3+);methanolate
Molecular Weight (g/mol) 214.862
PubChem CID 22289938
Formula Weight 214.8
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material Antimony(III) methoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'