missing translation for 'onlineSavingsMsg'
Learn More
Antimony(III) oxide, 99.6% min, Thermo Scientific Chemicals Product Code.: 11377317

Antimony(III) oxide, 99.6% min, Thermo Scientific Chemicals

Product Code. 11377317
50 g
Click to view available options
Quantity:
50 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11377317

Brand: Thermo Scientific Alfa Aesar 011579.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It finds applications in flame retardants, catalysts, batteries, and alloys. It acts as an opacifying agent in glasses, ceramics and enamels. It is used as a catalyst in the manufacture of PET plastic (polyethylene terephthalate), and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
Synonym Antimony trioxide
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1550°C (sublimation)
Melting Point 656°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.6% min (Metal basis)
Solubility Information Poor solubility in water. Slowly dissolves in warm HCl or warm KOH solutions.
Formula Weight 291.5
Odor Odorless
Appearance White powder
Density 5.67 g/cm3
Chemical Name or Material Antimony(III) oxide
Show More Show Less
compliance-icons
Product Identifier
  • Antimony(III) oxide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Antimony trioxide, Lead monoxide, Arsenic trioxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony(III) oxide, 99.6% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.