Learn More
Antimony(III) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
186.93 € - 754.76 €
Chemical Identifiers
CAS | 1309-64-4 |
---|---|
Molecular Formula | O3Sb2 |
Molecular Weight (g/mol) | 291.52 |
MDL Number | MFCD00011214 |
InChI Key | GHPGOEFPKIHBNM-UHFFFAOYSA-N |
IUPAC Name | diantimony(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Sb+3].[Sb+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11343398
|
Thermo Scientific Alfa Aesar
036314.18 |
50 g |
186.93 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333398
|
Thermo Scientific Alfa Aesar
036314.30 |
250 g |
754.76 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Flame retardant for wide range of plastics, rubbers, paper and textiles . Catalyst in PET production. Activator in glass industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1309-64-4 | |
291.52 | |
GHPGOEFPKIHBNM-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Sb+3].[Sb+3] |
O3Sb2 | |
MFCD00011214 | |
diantimony(3+) trioxidandiide |
Specifications
1550°C (sublimation) | |
1309-64-4 | |
Sintered Lump | |
O3Sb2 | |
Very-slightly soluble in cold water. Slightly soluble in hot water,dilute sulfuric acid,and nitric acid. Soluble in solutions of alkali hydroxide and sulfides,hydrochloric acid,tartaric acid,and acetic acid | |
[O--].[O--].[O--].[Sb+3].[Sb+3] | |
291.52 | |
Odorless | |
Antimony(III) oxide |
656°C | |
White | |
99.99% (Metals basis) | |
MFCD00011214 | |
GHPGOEFPKIHBNM-UHFFFAOYSA-N | |
diantimony(3+) trioxidandiide | |
291.5 | |
5.67 g/cm3 |
Safety and Handling
Do not breathe dust. Wear suitable protective clothing and gloves
P201-P202-P281-P308+P313-P501c
H351
missing translation for 'einecsNumber' : 215-175-0
missing translation for 'rtecsNumber' : CC5650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.