Learn More
Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 1309-64-4 |
---|---|
Molecular Formula | O3Sb2 |
Molecular Weight (g/mol) | 291.52 |
MDL Number | MFCD00011214 |
InChI Key | GHPGOEFPKIHBNM-UHFFFAOYSA-N |
IUPAC Name | diantimony(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Sb+3].[Sb+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11320839
|
Thermo Scientific Alfa Aesar
010641.06 |
5 g |
N/A
|
N/A | |||||
11330839
|
Thermo Scientific Alfa Aesar
010641.14 |
25 g |
N/A
|
N/A | |||||
15475955
|
Thermo Scientific Alfa Aesar
010641.18 |
50 g |
N/A
|
N/A | |||||
11340839
|
Thermo Scientific Alfa Aesar
010641.22 |
100 g |
N/A
|
N/A | |||||
Description
Antimony(III) oxide finds use as flame retardant wherein it synergistically acts with halogenated materials. It is employed in opacifying glasses, ceramics and enamels. In the preparation of optical and ruby glass, antimony trioxide serves as a bubble remover. Further, it is a useful catalyst in the production of polyethylene terephthalate (PET plastic) and the vulcanization of rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1309-64-4 | |
291.52 | |
GHPGOEFPKIHBNM-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Sb+3].[Sb+3] |
O3Sb2 | |
MFCD00011214 | |
diantimony(3+) trioxidandiide |
Specifications
1550°C (sublimation) | |
1309-64-4 | |
Powder | |
O3Sb2 | |
Very slightly soluble in water; soluble in potassium hydroxide,hydrochloricacid and acetic acid; insoluble in organic solvents. | |
[O--].[O--].[O--].[Sb+3].[Sb+3] | |
291.52 | |
Odorless | |
Antimony(III) oxide |
656°C | |
White | |
99.99% (Metals basis) | |
MFCD00011214 | |
GHPGOEFPKIHBNM-UHFFFAOYSA-N | |
diantimony(3+) trioxidandiide | |
291.5 | |
5.67 |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H351
missing translation for 'einecsNumber' : 215-175-0
missing translation for 'rtecsNumber' : CC5650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only