missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) selenide, 99.999% (metals basis), Thermo Scientific Chemicals

195.98 € - 788.93 €

Chemical Identifiers

CAS 1315-05-5
MDL Number MFCD00016319
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11332228
SDS View Documents
Thermo Scientific Alfa Aesar
013130.06
5 g
195.98 €
5g
Please to purchase this item. Need a web account? Register with us today!
11322228
SDS View Documents
Thermo Scientific Alfa Aesar
013130.14
25 g
788.93 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used to prepare large zintl ion, it is now actively explored for thin film solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1315-05-5
MFCD00016319

Specifications

611°C
Pieces
UN3077
Very slightly soluble in water
99.999%
(metals basis)
1315-05-5
MFCD00016319
14,712
480.38
Moisture sensitive
Antimony(III) selenide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H301-H330-H373

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

missing translation for 'einecsNumber' : 215-256-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.