Learn More
Antimony(III) sulfide, 98% (metals basis), Thermo Scientific Chemicals
65.10 € - 163.00 €
Chemical Identifiers
CAS | 1345-04-6 |
---|---|
Molecular Formula | S3Sb2 |
Molecular Weight (g/mol) | 339.70 |
MDL Number | MFCD00011217 |
InChI Key | NVWBARWTDVQPJD-UHFFFAOYSA-N |
Synonym | Antimony trisulfide |
IUPAC Name | diantimony(3+) trisulfanediide |
SMILES | [S--].[S--].[S--].[Sb+3].[Sb+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371849
|
Thermo Scientific Alfa Aesar
012521.30 |
250 g |
65.10 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361849
|
Thermo Scientific Alfa Aesar
012521.A1 |
1 kg |
163.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is manufactured for use in safety matches, military ammunition, explosives and fireworks. It also is used in the production of ruby-colored glass and in plastics as a flame retardant. percussion caps, camouflage paints, ruby glass
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1345-04-6 | |
339.70 | |
NVWBARWTDVQPJD-UHFFFAOYSA-N | |
diantimony(3+) trisulfanediide |
S3Sb2 | |
MFCD00011217 | |
Antimony trisulfide | |
[S--].[S--].[S--].[Sb+3].[Sb+3] |
Specifications
550°C | |
Black | |
98.0% min (Metal basis) | |
MFCD00011217 | |
Insoluble in water. Soluble in HCl with H2S evolution | |
[S--].[S--].[S--].[Sb+3].[Sb+3] | |
339.70 | |
Odorless | |
4.12-4.64g/cm3 |
1345-04-6 | |
Powder | |
S3Sb2 | |
Antimony trisulfide | |
NVWBARWTDVQPJD-UHFFFAOYSA-N | |
diantimony(3+) trisulfanediide | |
339.68 | |
Black powder | |
Antimony(III) sulfide |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H312-H315-H319-H335-H500
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
missing translation for 'einecsNumber' : 215-713-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only