missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) sulfide, 98%, Thermo Scientific Chemicals

36.28 € - 110.55 €

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
Synonym Antimony trisulfide
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10066143
missing translation for 'viewDocuments'
Thermo Scientific Acros
223591000
100 g Glass bottle
36.28 €
100g
Please to purchase this item. Need a web account? Register with us today!
10082930
missing translation for 'viewDocuments'
Thermo Scientific Acros
223595000
500 g Glass bottle
110.55 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1345-04-6
339.70
NVWBARWTDVQPJD-UHFFFAOYSA-N
diantimony(3+) trisulfanediide
S3Sb2
MFCD00011217
Antimony trisulfide
[S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

1150°C
1345-04-6
Crystalline Powder
S3Sb2
MFCD00011217
Antimony trisulfide
NVWBARWTDVQPJD-UHFFFAOYSA-N
diantimony(3+) trisulfanediide
339.67
Antimony(III) sulfide, 98%
550°C
Gray to Black
97.5% min. (Sb2S3)
Sb2S3
15, 701
Solubility in water: insoluble. Other solubilities: soluble in conc. hcl and alkali hydroxides,soluble in alcohol,soluble in amminium hydrogen sulfide,insoluble in acetic acid
[S--].[S--].[S--].[Sb+3].[Sb+3]
339.70
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 215-713-4

missing translation for 'rtecsNumber' : CC9450000

missing translation for 'tsca' : TSCA

Product Identifier
  • Antimony(III) sulfide

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.