missing translation for 'onlineSavingsMsg'
Learn More

Antimony pentafluoride, 99+%

Product Code. p-222634 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10016812

Brand: Thermo Scientific Acros 174430250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7783-70-2 | F5H8Sb | 224.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-70-2
Molecular Formula F5H8Sb
Molecular Weight (g/mol) 224.82
MDL Number MFCD00011219
InChI Key VBKPQOIVRXDIEL-UHFFFAOYSA-N
Synonym antimony pentafluoride, antimony v fluoride, pentafluoroantimony, antimony v pentafluoride, antimony fluoride sbf5, hsdb 442, antimonypentafluoride, pentafluorostibine, pentafluorostiborane
IUPAC Name antimony pentafluoride
SMILES F.F.F.F.F.[SbH3]

Specifications

Boiling Point 149.4°C
Melting Point 7.0°C
Color White
Physical Form Shiny Flakes or Powder
Quantity 25 g
Assay Percent Range 99+%
Linear Formula SbF5
Fieser 05,19; 06,23; 09,2
Merck Index 15, 687
Solubility Information Solubility in water: reacts with water. Other solubilities: forming hf and antimonyoxide, soluble in kf and liquid so2
Formula Weight 216.74
Percent Purity ≥99%
Packaging Plastic bottle
Specific Gravity 2.99
Density 2.9900g/mL
Chemical Name or Material Antimony pentafluoride, 99.5+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Antimony pentafluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.