Learn More
Antimony pentasulfide, pure, Thermo Scientific Chemicals
24.62 € - 198.99 €
Chemical Identifiers
CAS | 1315-04-4 |
---|---|
Molecular Formula | S5Sb2 |
Molecular Weight (g/mol) | 403.82 |
MDL Number | MFCD00016321 |
InChI Key | PPKVREKQVQREQD-UHFFFAOYSA-N |
Synonym | antimony pentasulfide, antimony v sulfide, antimony red, antimonic sulfide, antimonial saffron, antimony sulfide sb2s5, antimony sulfide golden, golden antimony sulfide, diantimony pentasulphide, goldschwefel |
PubChem CID | 16683083 |
IUPAC Name | [bis(sulfanylidene)-$l^{5}-stibanyl]sulfanyl-bis(sulfanylidene)-$l^{5}-stibane |
SMILES | S=[Sb](=S)S[Sb](=S)=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15265076
|
Thermo Scientific Acros
330410500 |
50 g |
24.62 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15274218
|
Thermo Scientific Acros
330412500 |
250 g |
63.62 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15264218
|
Thermo Scientific Acros
330410010 |
1 kg |
198.99 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1315-04-4 | |
403.82 | |
PPKVREKQVQREQD-UHFFFAOYSA-N | |
16683083 | |
S=[Sb](=S)S[Sb](=S)=S |
S5Sb2 | |
MFCD00016321 | |
antimony pentasulfide, antimony v sulfide, antimony red, antimonic sulfide, antimonial saffron, antimony sulfide sb2s5, antimony sulfide golden, golden antimony sulfide, diantimony pentasulphide, goldschwefel | |
[bis(sulfanylidene)-$l^{5}-stibanyl]sulfanyl-bis(sulfanylidene)-$l^{5}-stibane |
Specifications
1315-04-4 | |
Sb2S5 | |
15,688 | |
Solubility in water: insoluble. | |
S=[Sb](=S)S[Sb](=S)=S | |
403.82 | |
403.83 | |
Pure |
S5Sb2 | |
MFCD00016321 | |
antimony pentasulfide, antimony v sulfide, antimony red, antimonic sulfide, antimonial saffron, antimony sulfide sb2s5, antimony sulfide golden, golden antimony sulfide, diantimony pentasulphide, goldschwefel | |
PPKVREKQVQREQD-UHFFFAOYSA-N | |
[bis(sulfanylidene)-$l^{5}-stibanyl]sulfanyl-bis(sulfanylidene)-$l^{5}-stibane | |
16683083 | |
≥54% (Sb) | |
Antimony pentasulfide |
Safety and Handling
GHS H Statement
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-255-5
- Antimony pentasulfide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcoholresistant foam to extinguish.
- P391-Collect spillage.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Antimony pentasulfide
RUO – Research Use Only