missing translation for 'onlineSavingsMsg'
Learn More
Antimony powder, -200 mesh, 99.5% (metals basis), Thermo Scientific Chemicals Product Code.: 11381138

Antimony powder, -200 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11381138
2 kg
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11381138

Brand: Thermo Scientific Alfa Aesar 010099.A3

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes germanShow MoreShow Less
PubChem CID 5354495
ChEBI CHEBI:30304
SMILES [Sb]

Specifications

Color Gray
UN Number UN2871
Quantity 2 kg
IUPAC Name distibyne
Percent Purity 99.5%
Mesh Size −200 Mesh
Odor Odorless
Form Powder
Assay (metals basis)
Chemical Name or Material Antimony powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number62500180 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog Number010099.A3,010099.22,010099.09,010099.36
Lot NumberJ07X030 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number010099.A3,010099.22,010099.09,010099.36
Lot Number61301273 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010099.A3,010099.22,010099.09,010099.36
Lot NumberB28Y020 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010099.A3,010099.22,010099.09,010099.36
Lot NumberN06D038 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number010099.A3,010099.22,010099.09,010099.36
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony powder, -200 mesh, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.