missing translation for 'onlineSavingsMsg'
Learn More

Antimony trichloride, 99.5%, Thermo Scientific Chemicals

99.39 € - 755.00 €

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10104270
SDS View Documents
Thermo Scientific Acros
201685000
500 g Glass bottle
99.39 €
500g
Please to purchase this item. Need a web account? Register with us today!
10032790
SDS View Documents
Thermo Scientific Acros
201680020
2 kg Glass bottle
274.37 €
2kg
Please to purchase this item. Need a web account? Register with us today!
10428463
SDS View Documents
Thermo Scientific Acros
201680100
10 kg Plastic Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10025-91-9
228.11
FAPDDOBMIUGHIN-UHFFFAOYSA-K
24814
[Cl-].[Cl-].[Cl-].[Sb+3]
Cl3Sb
MFCD00011212
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
CHEBI:74856

Specifications

223°C
10025-91-9
Liquid
Cl3Sb
MFCD00011212
15, 696
Solubility in water: 100g/L (25°C). Other solubilities: soluble in diluted hcl,,soluble in chloroform (+/- 22%),soluble in alcohol,benzene,carbon disulfide,,acetone,tartaric acid,ccl4,insoluble in pyridine,quinoline,organic bases
[Cl-].[Cl-].[Cl-].[Sb+3]
228.11
CHEBI:74856
99.50%
73°C
Colorless
99.4% min. (Iodimetry)
SbCl3
08,19
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
FAPDDOBMIUGHIN-UHFFFAOYSA-K
antimony(3+) trichloride
24814
228.11
Antimony trichloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 233-047-2

Product Identifier
  • Antimony trichloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.