Learn More
Antimony trichloride, ACS reagent, Thermo Scientific Chemicals
32.36 € - 522.60 €
Chemical Identifiers
CAS | 10025-91-9 |
---|---|
Molecular Formula | Cl3Sb |
Molecular Weight (g/mol) | 228.11 |
MDL Number | MFCD00011212 |
InChI Key | FAPDDOBMIUGHIN-UHFFFAOYSA-K |
Synonym | antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony |
PubChem CID | 24814 |
ChEBI | CHEBI:74856 |
SMILES | [Cl-].[Cl-].[Cl-].[Sb+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10616241
|
Thermo Scientific Acros
401370050 |
5 g | Glass bottle |
32.36 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10430661
|
Thermo Scientific Acros
401371000 |
100 g | Glass bottle |
46.63 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10720862
|
Thermo Scientific Acros
401375000 |
500 g | Glass bottle |
149.75 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13418548
|
Thermo Scientific Acros
401370025 |
2.5 kg | Glass Bottle |
522.60 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-91-9 | |
228.11 | |
FAPDDOBMIUGHIN-UHFFFAOYSA-K | |
24814 | |
[Cl-].[Cl-].[Cl-].[Sb+3] |
Cl3Sb | |
MFCD00011212 | |
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony | |
CHEBI:74856 |
Specifications
223.0°C | |
10025-91-9 | |
Powder, Crystals and/or Chunks | |
SbCl3 | |
08,19 | |
antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony | |
FAPDDOBMIUGHIN-UHFFFAOYSA-K | |
trichlorostibane | |
24814 | |
228.11 | |
ACS Reagent |
73.0°C | |
White | |
Cl3Sb | |
MFCD00011212 | |
15,696 | |
Solubility in water: 100g/L (25°C). Other solubilities: soluble in diluted hcl,, soluble in chloroform (+/- 22%), soluble in alcohol, benzene, carbon disulfide,, acetone, tartaric acid, ccl4, insoluble in pyridine, quinoline, organic bases | |
[Cl-].[Cl-].[Cl-].[Sb+3] | |
228.11 | |
CHEBI:74856 | |
≥99.0% (Iodimetry) | |
Antimony trichloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 233-047-2
missing translation for 'rtecsNumber' : CC4900000
missing translation for 'tsca' : TSCA
- Antimony trichloride
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Antimony trichloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.