Learn More
Antimony trifluoride, 98%, Thermo Scientific Chemicals
20.70 € - 133.67 €
Chemical Identifiers
CAS | 7783-56-4 |
---|---|
Molecular Formula | F3Sb |
Molecular Weight (g/mol) | 178.75 |
MDL Number | MFCD00011218 |
InChI Key | GUNJVIDCYZYFGV-UHFFFAOYSA-K |
Synonym | antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438 |
PubChem CID | 24554 |
IUPAC Name | trifluorostibane |
SMILES | F[Sb](F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10685452
|
Thermo Scientific Acros
191240050 |
5 g | Plastic bottle |
20.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10002450
|
Thermo Scientific Acros
191241000 |
100 g | Plastic bottle |
48.14 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10791081
|
Thermo Scientific Acros
191245000 |
500 g | Plastic bottle |
133.67 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-56-4 | |
178.75 | |
GUNJVIDCYZYFGV-UHFFFAOYSA-K | |
24554 | |
F[Sb](F)F |
F3Sb | |
MFCD00011218 | |
antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438 | |
trifluorostibane |
Specifications
376.0°C | |
7783-56-4 | |
Crystalline Powder | |
F3Sb | |
MFCD00011218 | |
15, 697 | |
Solubility in water: 3850g/L (20°C). Other solubilities: decomposes in water, insoluble in nh3 | |
F[Sb](F)F | |
178.75 | |
178.75 | |
Antimony trifluoride, 98% |
292.0°C | |
White to Gray | |
97.5% min. (Iodimetry) | |
SbF3 | |
05,526; 06,553 | |
antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438 | |
GUNJVIDCYZYFGV-UHFFFAOYSA-K | |
trifluorostibane | |
24554 | |
98% |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 232-009-2
missing translation for 'rtecsNumber' : CC5150000
missing translation for 'tsca' : TSCA
- Antimony trifluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only