missing translation for 'onlineSavingsMsg'
Learn More

Antimony Trioxide, 99+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10385500
Click to view available options
500g, Plastic powder jar

Specifications

Boiling Point 1550°C
Melting Point 656°C
CAS 1309-64-4
Color White
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula O3Sb2
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]
IUPAC Name diantimony(3+) trioxidandiide
Molecular Weight (g/mol) 291.52
Formula Weight 291.42g/mol
Packaging Plastic powder jar
Vapor Pressure 1.3 hPa 574
Show More Show Less
compliance-icons
Product Identifier
  • Antimony(III) oxide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.