missing translation for 'onlineSavingsMsg'
Learn More

Antimony(V) chloride, 98%, Thermo Scientific Chemicals

44.20 € - 432.00 €

Chemical Identifiers

CAS 7647-18-9
Molecular Formula Cl5Sb
Molecular Weight (g/mol) 299.02
MDL Number MFCD00011213
InChI Key VMPVEPPRYRXYNP-UHFFFAOYSA-I
Synonym antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
PubChem CID 24294
IUPAC Name antimony pentachloride
SMILES Cl[Sb](Cl)(Cl)(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394967
SDS View Documents
Thermo Scientific Alfa Aesar
041978.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11314977
SDS View Documents
Thermo Scientific Alfa Aesar
041978.30
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11304977
SDS View Documents
Thermo Scientific Alfa Aesar
041978.A1
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Antimony(V) chloride is used as a catalyst and an analytical reagent for testing alkaloids and cesium. It is also used as a polymerization catalyst as well as involved in the chlorination of organic compounds. It acts as a Lewis acid and a strong oxidizing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7647-18-9
299.02
VMPVEPPRYRXYNP-UHFFFAOYSA-I
24294
Cl[Sb](Cl)(Cl)(Cl)Cl
Cl5Sb
MFCD00011213
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
antimony pentachloride

Specifications

79°C (22 mm)
7647-18-9
Liquid
Cl5Sb
antimony v chloride, antimony pentachloride, pentachloroantimony, antimony perchloride, antimony chloride sbcl5, antimonpentachlorid, antimoonpentachloride, perchlorure d'antimoine, pentachlorure d'antimoine, antimonpentachlorid german
VMPVEPPRYRXYNP-UHFFFAOYSA-I
antimony pentachloride
24294
Pungent
Moisture sensitive
2.336 g/cm3
2.8°C
Red-Yellow
98%
MFCD00011213
Soluble in HCl,chloroform,carbon tetrachlorideMiscible with chloroform,hydrochloric acid,tartaric acid,carbon disulfide and carbon tetrachloride.
Cl[Sb](Cl)(Cl)(Cl)Cl
299.02
299.02
1.601
Red-yellow liquid
Antimony(V) chloride
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c

H302-H314-H335

missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II

missing translation for 'einecsNumber' : 231-601-8

missing translation for 'rtecsNumber' : CC5075000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.